Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCCCGCGCGGGCACCAGCCCC[C/G]GCGGCAGCCCGCCCGCGCGTCCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601181 | ||||||||||||||||||||
Literature Links: |
RANBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RANBP2 - RAN binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006267.4 | 496 | Intron | NP_006258.3 | |||
XM_005264002.2 | 496 | Intron | XP_005264059.1 | |||
XM_005264003.2 | 496 | Intron | XP_005264060.1 | |||
XM_005264004.2 | 496 | Intron | XP_005264061.1 | |||
XM_005264005.4 | 496 | Intron | XP_005264062.1 | |||
XM_005264007.2 | 496 | Intron | XP_005264064.1 | |||
XM_011511575.2 | 496 | Intron | XP_011509877.1 | |||
XM_011511576.2 | 496 | Intron | XP_011509878.1 | |||
XM_011511578.2 | 496 | Intron | XP_011509880.1 | |||
XM_017004623.1 | 496 | Intron | XP_016860112.1 | |||
XM_017004624.1 | 496 | Intron | XP_016860113.1 | |||
XM_017004625.1 | 496 | Intron | XP_016860114.1 |
SH3RF3 - SH3 domain containing ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099289.2 | 496 | Missense Mutation | CGC,GGC | R,G 132 | NP_001092759.1 | |
XM_011511109.2 | 496 | Missense Mutation | CGC,GGC | R,G 132 | XP_011509411.1 | |
XM_011511110.2 | 496 | Intron | XP_011509412.1 |
SH3RF3-AS1 - SH3RF3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |