Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTTTGACACTTGTTTTTGGGAC[A/T]GTCCGCTCTCAGCAGCTCTCCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM127 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM127 - transmembrane protein 127 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193304.2 | 963 | UTR 3 | NP_001180233.1 | |||
NM_017849.3 | 963 | UTR 3 | NP_060319.1 | |||
XM_017004450.1 | 963 | Missense Mutation | CAG,CTG | Q,L 48 | XP_016859939.1 | |
XM_017004451.1 | 963 | UTR 3 | XP_016859940.1 | |||
XM_017004452.1 | 963 | UTR 3 | XP_016859941.1 |