Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAAGGAAATAATAGGACTAGTAG[C/T]CCATGGTGTATGCAGATGTTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF2 - zinc finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017396.2 | Intron | NP_001017396.1 | ||||
NM_001282398.1 | Intron | NP_001269327.1 | ||||
NM_001291604.1 | Intron | NP_001278533.1 | ||||
NM_001291605.1 | Intron | NP_001278534.1 | ||||
NM_021088.3 | Intron | NP_066574.2 |