Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCTGACTGTGAAGAAAAGCTGG[G/T]GTGGAAAGAGAGGCCATGGCCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1R1 - interleukin 1 receptor type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000877.3 | Intron | NP_000868.1 | ||||
NM_001288706.1 | Intron | NP_001275635.1 | ||||
NM_001320978.1 | Intron | NP_001307907.1 | ||||
NM_001320980.1 | Intron | NP_001307909.1 | ||||
NM_001320981.1 | Intron | NP_001307910.1 | ||||
NM_001320982.1 | Intron | NP_001307911.1 | ||||
NM_001320983.1 | Intron | NP_001307912.1 | ||||
NM_001320984.1 | Intron | NP_001307913.1 | ||||
NM_001320985.1 | Intron | NP_001307914.1 | ||||
NM_001320986.1 | Intron | NP_001307915.1 | ||||
XM_005263930.4 | Intron | XP_005263987.1 | ||||
XM_005263934.4 | Intron | XP_005263991.1 | ||||
XM_011511115.2 | Intron | XP_011509417.1 | ||||
XM_011511116.1 | Intron | XP_011509418.1 | ||||
XM_011511117.1 | Intron | XP_011509419.1 | ||||
XM_011511118.2 | Intron | XP_011509420.1 | ||||
XM_017003989.1 | Intron | XP_016859478.1 |