Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGATTGCGTAAACTTAAACATA[C/T]CACTTTACAGTAAAACCTAAAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPAST - spastin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014946.3 | Intron | NP_055761.2 | ||||
NM_199436.1 | Intron | NP_955468.1 | ||||
XM_005264516.4 | Intron | XP_005264573.1 | ||||
XM_011533067.2 | Intron | XP_011531369.1 | ||||
XM_017004777.1 | Intron | XP_016860266.1 | ||||
XM_017004778.1 | Intron | XP_016860267.1 |