Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAAGATGATGATCCTGAAGATAA[A/C]AAGCCACTCTTGTATGTAAAAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
YIPF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
YIPF4 - Yip1 domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032312.3 | 489 | Silent Mutation | AAA,AAC | K,N 74 | NP_115688.1 | |
XM_005264599.3 | 489 | Missense Mutation | AAA,AAC | K,N 74 | XP_005264656.1 | |
XM_011533125.2 | 489 | Missense Mutation | AAA,AAC | K,N 74 | XP_011531427.1 |