Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTTTTTAATCCATTCTTCTTT[C/T]GATGTGCATTATGGTTGTTCCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2AK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2AK2 - eukaryotic translation initiation factor 2 alpha kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135651.2 | 2784 | UTR 3 | NP_001129123.1 | |||
NM_001135652.2 | 2784 | UTR 3 | NP_001129124.1 | |||
NM_002759.3 | 2784 | UTR 3 | NP_002750.1 | |||
XM_011532987.2 | 2784 | UTR 3 | XP_011531289.1 | |||
XM_017004503.1 | 2784 | UTR 3 | XP_016859992.1 |
GPATCH11 - G-patch domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |