Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGGTGGGGCATAATATTTTGG[A/G]ATGGCAAGTGCCAAAAAATTTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601771 MIM: 611872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP1B1 - cytochrome P450 family 1 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000104.3 | 4193 | UTR 3 | NP_000095.2 |
RMDN2 - regulator of microtubule dynamics 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170791.2 | 4193 | Intron | NP_001164262.1 | |||
NM_001170792.2 | 4193 | Intron | NP_001164263.1 | |||
NM_001170793.2 | 4193 | Intron | NP_001164264.1 | |||
NM_001322211.1 | 4193 | Intron | NP_001309140.1 | |||
NM_001322212.1 | 4193 | Intron | NP_001309141.1 | |||
NM_144713.4 | 4193 | Intron | NP_653314.3 | |||
XM_011532613.2 | 4193 | Intron | XP_011530915.1 | |||
XM_011532614.2 | 4193 | Intron | XP_011530916.1 | |||
XM_011532615.2 | 4193 | Intron | XP_011530917.2 | |||
XM_011532616.2 | 4193 | Intron | XP_011530918.1 | |||
XM_011532617.2 | 4193 | Intron | XP_011530919.1 | |||
XM_011532618.2 | 4193 | Intron | XP_011530920.1 | |||
XM_011532619.2 | 4193 | Intron | XP_011530921.1 | |||
XM_017003474.1 | 4193 | Intron | XP_016858963.1 | |||
XM_017003475.1 | 4193 | Intron | XP_016858964.1 | |||
XM_017003476.1 | 4193 | Intron | XP_016858965.1 | |||
XM_017003477.1 | 4193 | Intron | XP_016858966.1 | |||
XM_017003478.1 | 4193 | Intron | XP_016858967.1 | |||
XM_017003479.1 | 4193 | Intron | XP_016858968.1 |