Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAGCCTGAAAGCTGCAATTAGT[A/G]TAAAGATCATCCAGAAATGTTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137030 MIM: 600572 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALM - galactose mutarotase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRSF7 - serine and arginine rich splicing factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031684.2 | 1607 | UTR 3 | NP_001026854.1 | |||
NM_001195446.1 | 1607 | UTR 3 | NP_001182375.1 | |||
XM_005264484.2 | 1607 | Intron | XP_005264541.1 | |||
XM_005264485.2 | 1607 | Intron | XP_005264542.1 | |||
XM_011533032.2 | 1607 | Intron | XP_011531334.1 |