Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTCAAAGAAGAAAAATATTGA[A/G]TAAAGTCATCATGTTCCCTTATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF33 - Rho guanine nucleotide exchange factor 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOS1 - SOS Ras/Rac guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005633.3 | 7458 | UTR 3 | NP_005624.2 | |||
XM_005264515.4 | 7458 | Intron | XP_005264572.1 | |||
XM_011533062.2 | 7458 | Intron | XP_011531364.1 | |||
XM_011533064.2 | 7458 | Intron | XP_011531366.1 | |||
XM_011533066.2 | 7458 | Intron | XP_011531368.1 |