Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGGAAGATGCTCCAGAGAGAC[A/G]AGGCTGCGGCGGAGGAGGTGGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPM1B - protein phosphatase, Mg2+/Mn2+ dependent 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033557.2 | 227 | UTR 5 | NP_001028729.1 | |||
NM_002706.5 | 227 | UTR 5 | NP_002697.1 | |||
NM_177968.3 | 227 | UTR 5 | NP_808907.1 | |||
NM_177969.3 | 227 | UTR 5 | NP_808908.1 | |||
XM_011532936.2 | 227 | UTR 5 | XP_011531238.1 | |||
XM_017004395.1 | 227 | UTR 5 | XP_016859884.1 | |||
XM_017004396.1 | 227 | UTR 5 | XP_016859885.1 | |||
XM_017004397.1 | 227 | UTR 5 | XP_016859886.1 |