Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAACCAATGCCATATTTTATTT[C/T]CAAAATTATTAGACTTCAACAGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf61 - chromosome 2 open reading frame 61 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CALM2 - calmodulin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305624.1 | 1338 | UTR 3 | NP_001292553.1 | |||
NM_001305625.1 | 1338 | UTR 3 | NP_001292554.1 | |||
NM_001305626.1 | 1338 | UTR 3 | NP_001292555.1 | |||
NM_001743.5 | 1338 | UTR 3 | NP_001734.1 |