Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACGGGGCGGGGGAGGCCATGGCC[C/T]CGGCTGAGTTGCAGGGGAAGTACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PPP1R21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R21 - protein phosphatase 1 regulatory subunit 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135629.2 | 192 | Missense Mutation | CCG,TCG | P,S 3 | NP_001129101.1 | |
NM_001193475.1 | 192 | Missense Mutation | CCG,TCG | P,S 3 | NP_001180404.1 | |
NM_152994.4 | 192 | Missense Mutation | CCG,TCG | P,S 3 | NP_694539.1 | |
XM_017003301.1 | 192 | UTR 5 | XP_016858790.1 | |||
XM_017003302.1 | 192 | UTR 5 | XP_016858791.1 |