Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCGTTAGTCAGAAGCTGTTAAG[C/T]AACGAGATGGCCACGGTCCACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605760 MIM: 611229 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB3 - ankyrin repeat and SOCS box containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201965.1 | 333 | UTR 5 | NP_001188894.1 | |||
NM_016115.4 | 333 | Intron | NP_057199.1 | |||
NM_145863.2 | 333 | Intron | NP_665862.1 |
ERLEC1 - endoplasmic reticulum lectin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127397.2 | 333 | Intron | NP_001120869.1 | |||
NM_001127398.2 | 333 | Intron | NP_001120870.1 | |||
NM_015701.4 | 333 | Intron | NP_056516.2 | |||
XM_011532766.1 | 333 | Intron | XP_011531068.1 |
GPR75-ASB3 - GPR75-ASB3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164165.1 | 333 | Intron | NP_001157637.1 |