Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGAAAATCGATGAGCTTGTGGA[C/T]GAGCTGGAAACAGCCCCATGAAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRED2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRED2 - sprouty related EVH1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128210.1 | 2899 | UTR 3 | NP_001121682.1 | |||
NM_181784.2 | 2899 | UTR 3 | NP_861449.2 | |||
XM_005264200.4 | 2899 | Intron | XP_005264257.2 | |||
XM_005264201.1 | 2899 | UTR 3 | XP_005264258.1 | |||
XM_005264202.4 | 2899 | Intron | XP_005264259.1 | |||
XM_017003576.1 | 2899 | UTR 3 | XP_016859065.1 |