Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
AGCCACTAAAACTGGGATCACAAAC[A/G]CCCTAGCCTTGTGTTCATTCTTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601739 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEIS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEIS1 - Meis homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002398.2 | Intron | NP_002389.1 | ||||
XM_005264321.2 | Intron | XP_005264378.1 | ||||
XM_005264322.2 | Intron | XP_005264379.1 | ||||
XM_005264323.2 | Intron | XP_005264380.1 | ||||
XM_005264324.4 | Intron | XP_005264381.1 | ||||
XM_005264325.4 | Intron | XP_005264382.1 | ||||
XM_017004141.1 | Intron | XP_016859630.1 | ||||
XM_017004142.1 | Intron | XP_016859631.1 | ||||
XM_017004143.1 | Intron | XP_016859632.1 | ||||
XM_017004144.1 | Intron | XP_016859633.1 | ||||
XM_017004145.1 | Intron | XP_016859634.1 | ||||
XM_017004146.1 | Intron | XP_016859635.1 |
MEIS1-AS2 - MEIS1 antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEIS1-AS3 - MEIS1 antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |