Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATGCAGGTAAAGACAACACCA[A/G]AAAGTATATAGACTAAGTAGATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GMCL1 - germ cell-less, spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178439.4 | Intron | NP_848526.1 | ||||
XM_011533033.1 | Intron | XP_011531335.1 | ||||
XM_011533034.2 | Intron | XP_011531336.1 | ||||
XM_017004704.1 | Intron | XP_016860193.1 | ||||
XM_017004705.1 | Intron | XP_016860194.1 |