Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGTTTTTCTATACGACCGTAG[C/G]TTTCTGCTATGCTTTCTACTTCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf42 - chromosome 2 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017880.1 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | NP_060350.1 | |
XM_005264389.2 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_005264446.1 | |
XM_005264390.2 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_005264447.1 | |
XM_005264391.2 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_005264448.1 | |
XM_011532937.2 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_011531239.1 | |
XM_017004398.1 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_016859887.1 | |
XM_017004399.1 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_016859888.1 | |
XM_017004400.1 | 1972 | Missense Mutation | ACC,AGC | T,S 484 | XP_016859889.1 |