Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACGCAGTATCCTAGGATCCTCT[C/T]GGACATCAGACCATTTACCTGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGUOK-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGUOK-AS1 - DGUOK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TET3 - tet methylcytosine dioxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287491.1 | Intron | NP_001274420.1 | ||||
XM_005264187.3 | Intron | XP_005264244.1 | ||||
XM_011532682.2 | Intron | XP_011530984.1 | ||||
XM_011532683.2 | Intron | XP_011530985.1 | ||||
XM_011532684.2 | Intron | XP_011530986.1 | ||||
XM_011532685.2 | Intron | XP_011530987.1 | ||||
XM_011532686.2 | Intron | XP_011530988.1 | ||||
XM_011532687.2 | Intron | XP_011530989.1 | ||||
XM_011532688.2 | Intron | XP_011530990.1 | ||||
XM_011532689.2 | Intron | XP_011530991.1 | ||||
XM_011532690.1 | Intron | XP_011530992.1 | ||||
XM_017003566.1 | Intron | XP_016859055.1 | ||||
XM_017003567.1 | Intron | XP_016859056.1 |