Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGAGCGCGCGCGTCAAGTCCGC[A/G]GAAAGTTTTACTAGTCAGCTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607164 MIM: 610231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LBX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LBX2 - ladybird homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009812.1 | 190 | Intron | NP_001009812.1 | |||
NM_001282430.1 | 190 | Missense Mutation | CGC,TGC | R,C 57 | NP_001269359.1 |
LBX2-AS1 - LBX2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCGF1 - polycomb group ring finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC31 - tetratricopeptide repeat domain 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |