Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACAGGGCGAGGAGGAGGAGCAG[T/C]AGCAGGAGCAGCAACAGTTTGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615288 MIM: 603681 | ||||||||||||||||||||
Literature Links: |
DRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRC1 - dynein regulatory complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOF - otoferlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287489.1 | 4001 | Intron | NP_001274418.1 | |||
NM_004802.3 | 4001 | Silent Mutation | CTA,CTG | L,L 1207 | NP_004793.2 | |
NM_194248.2 | 4001 | Silent Mutation | CTA,CTG | L,L 1974 | NP_919224.1 | |
NM_194322.2 | 4001 | Silent Mutation | CTA,CTG | L,L 1284 | NP_919303.1 | |
NM_194323.2 | 4001 | Intron | NP_919304.1 | |||
XM_017005338.1 | 4001 | Silent Mutation | CTA,CTG | L,L 1954 | XP_016860827.1 |