Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCAGATGAAGCTCGTGTCGGGC[T/C]GGCTGGAGTATGAAGGGTAGAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615288 MIM: 603681 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRC1 - dynein regulatory complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOF - otoferlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287489.1 | 3895 | Intron | NP_001274418.1 | |||
NM_004802.3 | 3895 | Missense Mutation | NP_004793.2 | |||
NM_194248.2 | 3895 | Missense Mutation | NP_919224.1 | |||
NM_194322.2 | 3895 | Missense Mutation | NP_919303.1 | |||
NM_194323.2 | 3895 | Intron | NP_919304.1 | |||
XM_017005338.1 | 3895 | Missense Mutation | XP_016860827.1 |