Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTAATCATTTATCAAACATGAA[A/G]TCTGGTGCATTAAAGGAATATCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604652 MIM: 603062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCF7L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCF7L1 - transcription factor 7 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGOLN2 - trans-golgi network protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206840.1 | 5910 | UTR 3 | NP_001193769.1 | |||
NM_001206841.1 | 5910 | UTR 3 | NP_001193770.1 | |||
NM_001206844.1 | 5910 | UTR 3 | NP_001193773.1 | |||
NM_006464.3 | 5910 | UTR 3 | NP_006455.2 |