Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCTCCTCATACTGTTCAGTCA[C/T]GGTTCTCATGGCTCCATAGTGTTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609777 | |||||||||||||||||||||||
Literature Links: |
XIRP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
XIRP1 - xin actin binding repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198621.2 | 5106 | UTR 3 | NP_001185550.1 | |||
NM_194293.2 | 5106 | Missense Mutation | NP_919269.2 | |||
XM_005264909.3 | 5106 | Missense Mutation | XP_005264966.1 |