Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTGCTCTCTATGGACATTAAC[A/G]CAGTACACAAATATCCGTGCTCAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607939 | |||||||||||||||||||||||
Literature Links: |
SUMF1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SUMF1 - sulfatase modifying factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164674.1 | 1637 | Intron | NP_001158146.1 | |||
NM_001164675.1 | 1637 | Intron | NP_001158147.1 | |||
NM_182760.3 | 1637 | Intron | NP_877437.2 | |||
XM_011533624.2 | 1637 | UTR 3 | XP_011531926.1 | |||
XM_011533625.2 | 1637 | Intron | XP_011531927.1 | |||
XM_011533626.2 | 1637 | Intron | XP_011531928.1 | |||
XM_017006252.1 | 1637 | UTR 3 | XP_016861741.1 | |||
XM_017006253.1 | 1637 | UTR 3 | XP_016861742.1 | |||
XM_017006254.1 | 1637 | Intron | XP_016861743.1 | |||
XM_017006255.1 | 1637 | Intron | XP_016861744.1 |