Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCATACTCCTCGGCACTGCGAC[A/G]GCCAAAATCCATCCAGCCCATGTAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 118440 | |||||||||||||||||||||||
Literature Links: |
CCK PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCK - cholecystokinin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000729.5 | 1207 | Missense Mutation | CGT,TGT | R,C 105 | NP_000720.1 | |
NM_001174138.2 | 1207 | Missense Mutation | CGT,TGT | R,C 105 | NP_001167609.1 |