Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCATTTGTCCCTCTACTTTGTT[C/T]TATTTATAAGCTCCATCGAGTCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604028 MIM: 606473 | |||||||||||||||||||||||
Literature Links: |
SEC22C PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SEC22C - SEC22 homolog C, vesicle trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201572.1 | Intron | NP_001188501.1 | ||||
NM_001201584.1 | Intron | NP_001188513.1 | ||||
NM_004206.3 | Intron | NP_004197.1 | ||||
NM_032970.3 | Intron | NP_116752.1 | ||||
XM_005265533.1 | Intron | XP_005265590.1 | ||||
XM_005265534.1 | Intron | XP_005265591.1 | ||||
XM_005265535.1 | Intron | XP_005265592.1 | ||||
XM_011534212.1 | Intron | XP_011532514.1 | ||||
XM_017007438.1 | Intron | XP_016862927.1 | ||||
XM_017007439.1 | Intron | XP_016862928.1 |
SS18L2 - SS18 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016305.3 | Intron | NP_057389.1 | ||||
XM_017006548.1 | Intron | XP_016862037.1 | ||||
XM_017006549.1 | Intron | XP_016862038.1 |