Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTGCCCTTAGAATGAGTCCACT[A/G]AGCAATGTGAAGTTGGCGGTGTCCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611735 | |||||||||||||||||||||||
Literature Links: |
CDCP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CDCP1 - CUB domain containing protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022842.4 | 2795 | UTR 3 | NP_073753.3 | |||
NM_178181.2 | 2795 | Intron | NP_835488.1 | |||
XM_011534024.1 | 2795 | UTR 3 | XP_011532326.1 | |||
XM_017007070.1 | 2795 | UTR 3 | XP_016862559.1 |