Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATCCTCCAGGATTCAGAGCAG[A/G]CTCGGTGCTGTGTTTGCCATTGAGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605616 | |||||||||||||||||||||||
Literature Links: |
SLC6A20 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC6A20 - solute carrier family 6 member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020208.3 | 4864 | UTR 3 | NP_064593.1 | |||
NM_022405.3 | 4864 | UTR 3 | NP_071800.1 | |||
XM_011533847.2 | 4864 | UTR 3 | XP_011532149.1 | |||
XM_011533848.2 | 4864 | Intron | XP_011532150.1 |