Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGCCTGTGCAGCCAGCACACC[C/T]TGGGCCTTGCGCTCAGCTAGGGCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 150325 MIM: 614471 | |||||||||||||||||||||||
Literature Links: |
LAMB2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LAMB2 - laminin subunit beta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002292.3 | 5264 | Silent Mutation | CAA,CAG | Q,Q 1722 | NP_002283.3 | |
XM_005265127.3 | 5264 | Silent Mutation | CAA,CAG | Q,Q 1722 | XP_005265184.1 |