Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATCTTCAGGTGGACAGAGGCCC[A/G]GTGAGGACTGTGACTAAGCGGGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607726 | ||||||||||||||||||||
Literature Links: |
PARP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARP3 - poly(ADP-ribose) polymerase family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003931.3 | 927 | Silent Mutation | CCA,CCG | P,P 179 | NP_001003931.3 | |
NM_005485.5 | 927 | Silent Mutation | CCA,CCG | P,P 172 | NP_005476.4 | |
XM_005264779.4 | 927 | Silent Mutation | CCA,CCG | P,P 172 | XP_005264836.2 | |
XM_017005490.1 | 927 | Silent Mutation | CCA,CCG | P,P 172 | XP_016860979.1 |
RRP9 - ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |