Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCACTGGGGGAGGTGACCTGAA[C/T]AAGAGGCATAGTATAAAGCCACAAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616966 | |||||||||||||||||||||||
Literature Links: |
ABHD6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ABHD6 - abhydrolase domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320126.1 | Intron | NP_001307055.1 | ||||
NM_020676.6 | Intron | NP_065727.4 | ||||
XM_005265334.3 | Intron | XP_005265391.1 | ||||
XM_005265335.3 | Intron | XP_005265392.1 | ||||
XM_017006930.1 | Intron | XP_016862419.1 |