Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACATTACAAAACAAGGTTTCAC[A/G]ATCCGCAACACCCGGGTGCATCACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601641 | |||||||||||||||||||||||
Literature Links: |
ACOX2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACOX2 - acyl-CoA oxidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCTD6 - potassium channel tetramerization domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128214.1 | 2061 | Silent Mutation | ACA,ACG | T,T 204 | NP_001121686.1 | |
NM_153331.3 | 2061 | Silent Mutation | ACA,ACG | T,T 204 | NP_699162.3 | |
XM_005264937.2 | 2061 | Silent Mutation | ACA,ACG | T,T 204 | XP_005264994.1 |