Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGCTGGGGAGTAAGGAGCCAAG[A/G]ACATGCCAGTGTGGTTCCTGGGCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CPNE9 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CPNE9 - copine family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308388.1 | Intron | NP_001295317.1 | ||||
NM_153635.2 | Intron | NP_705899.2 | ||||
XM_011533386.2 | Intron | XP_011531688.1 | ||||
XM_011533388.2 | Intron | XP_011531690.1 | ||||
XM_011533389.2 | Intron | XP_011531691.1 | ||||
XM_017005747.1 | Intron | XP_016861236.1 |