Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGTGCCTGGGAGAAGGAGCC[A/G]ATGTAGGTGGAGGTGCAAGGGCAGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ARPC4-TTLL3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARPC4-TTLL3 - ARPC4-TTLL3 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPUSD3 - RNA pseudouridylate synthase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142547.1 | 808 | Missense Mutation | CGG,TGG | R,W 301 | NP_001136019.1 | |
NM_173659.3 | 808 | Missense Mutation | CGG,TGG | R,W 316 | NP_775930.2 | |
XM_011533627.2 | 808 | UTR 3 | XP_011531929.1 |
TTLL3 - tubulin tyrosine ligase like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |