Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGAATATAGACATAACTACAA[C/T]GCTGATATTATTTCTTGTTTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601695 MIM: 607241 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSN3 - casein kappa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005212.2 | Intron | NP_005203.2 | ||||
XM_011531620.2 | Intron | XP_011529922.1 | ||||
XM_017007761.1 | Intron | XP_016863250.1 |
FDCSP - follicular dendritic cell secreted protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |