Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGAGACCACAATATTCAGCAAT[C/G]AGCTCCCTGGTATTTACATACTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRSF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRSF1 - G-rich RNA sequence binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098477.1 | 3605 | UTR 3 | NP_001091947.1 | |||
NM_002092.3 | 3605 | UTR 3 | NP_002083.3 | |||
XM_005265681.3 | 3605 | Intron | XP_005265738.1 | |||
XM_011531897.2 | 3605 | Intron | XP_011530199.1 |