Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCCGATGTAATATGTCTATGTGA[C/T]ATAAACATTTTTTTGAAAAATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASSF6 - Ras association domain family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270391.1 | 4600 | UTR 3 | NP_001257320.1 | |||
NM_001270392.1 | 4600 | UTR 3 | NP_001257321.1 | |||
NM_177532.4 | 4600 | UTR 3 | NP_803876.1 | |||
NM_201431.2 | 4600 | UTR 3 | NP_958834.1 | |||
XM_011531687.2 | 4600 | Intron | XP_011529989.1 | |||
XM_011531688.1 | 4600 | UTR 3 | XP_011529990.1 | |||
XM_011531689.2 | 4600 | UTR 3 | XP_011529991.1 | |||
XM_011531690.2 | 4600 | UTR 3 | XP_011529992.1 | |||
XM_011531691.1 | 4600 | UTR 3 | XP_011529993.1 | |||
XM_017007837.1 | 4600 | UTR 3 | XP_016863326.1 | |||
XM_017007838.1 | 4600 | Intron | XP_016863327.1 |