Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACAGCGGGGCTTGCAGGTCCAAG[A/C]AAATTTTCCTTCCATTCTTCAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173460 MIM: 121010 | ||||||||||||||||||||
Literature Links: |
PF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PF4 - platelet factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002619.3 | 419 | Missense Mutation | TGC,TTC | C,F 83 | NP_002610.1 | |
XM_005265696.3 | 419 | Intron | XP_005265753.1 |
PPBP - pro-platelet basic protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |