Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAACTTACATATTCCATCATG[C/T]TATTCGTTTCACATTTCCTTTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604332 | ||||||||||||||||||||
Literature Links: |
CHIC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHIC2 - cysteine rich hydrophobic domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012110.3 | 698 | Missense Mutation | AAC,AGC | N,S 145 | NP_036242.1 | |
XM_006714037.3 | 698 | Missense Mutation | AAC,AGC | N,S 117 | XP_006714100.1 | |
XM_011534382.2 | 698 | Intron | XP_011532684.1 |