Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAACAAGTAAATTGTGTAATAC[G/A]TCAGATGGTGATAGGTTCTGTAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCUN1D4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCUN1D4 - defective in cullin neddylation 1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040402.2 | Intron | NP_001035492.1 | ||||
NM_001287755.1 | Intron | NP_001274684.1 | ||||
NM_001287757.1 | Intron | NP_001274686.1 | ||||
NM_015115.3 | Intron | NP_055930.2 | ||||
XM_005265731.3 | Intron | XP_005265788.1 | ||||
XM_011534379.2 | Intron | XP_011532681.1 | ||||
XM_011534380.1 | Intron | XP_011532682.1 | ||||
XM_017007912.1 | Intron | XP_016863401.1 | ||||
XM_017007913.1 | Intron | XP_016863402.1 |