Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATCCCAAGGAGGGGGCGATGAT[A/C]ATCAGTAGTTGGGAGATGGGCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610096 | ||||||||||||||||||||
Literature Links: |
TIMD4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TIMD4 - T-cell immunoglobulin and mucin domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146726.1 | 924 | Missense Mutation | ATG,ATT | M,I 289 | NP_001140198.1 | |
NM_138379.2 | 924 | Missense Mutation | ATG,ATT | M,I 317 | NP_612388.2 | |
XM_011534694.2 | 924 | Intron | XP_011532996.1 | |||
XM_017010021.1 | 924 | Missense Mutation | ATG,ATT | M,I 262 | XP_016865510.1 |