Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAAAGACAAATATTGGTGCTAA[C/T]ATGTCTTTATTACCTCTCCAACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606323 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYFIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYFIP2 - cytoplasmic FMR1 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037333.2 | 1684 | Intron | NP_001032410.1 | |||
NM_001291721.1 | 1684 | Intron | NP_001278650.1 | |||
NM_001291722.1 | 1684 | Intron | NP_001278651.1 | |||
NM_014376.3 | 1684 | Intron | NP_055191.2 | |||
XM_011534516.2 | 1684 | Intron | XP_011532818.1 | |||
XM_017009341.1 | 1684 | Intron | XP_016864830.1 | |||
XM_017009342.1 | 1684 | Intron | XP_016864831.1 |
FNDC9 - fibronectin type III domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001343.3 | 1684 | UTR 3 | NP_001001343.2 |