Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTAGAAGGGTGTACCAGCGTC[C/T]CTTTGTTCAGAGCAGCATTGAAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIMC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIMC1 - SUMO interacting motifs containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308195.1 | Intron | NP_001295124.1 | ||||
NM_001308196.1 | Intron | NP_001295125.1 | ||||
NM_001308200.1 | Intron | NP_001295129.1 | ||||
NM_198567.5 | Intron | NP_940969.3 | ||||
XM_011534553.2 | Intron | XP_011532855.1 | ||||
XM_011534554.2 | Intron | XP_011532856.1 | ||||
XM_011534556.2 | Intron | XP_011532858.1 | ||||
XM_017009454.1 | Intron | XP_016864943.1 | ||||
XM_017009455.1 | Intron | XP_016864944.1 | ||||
XM_017009456.1 | Intron | XP_016864945.1 |