Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGTCACTGAAGCAGACCACACA[C/T]CTGTGGAGGGGCGCGGAGCGTCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF44 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF44 - ring finger protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014901.4 | 1553 | Silent Mutation | CTA,CTG | L,L 379 | NP_055716.1 | |
XM_005265840.2 | 1553 | Silent Mutation | CTA,CTG | L,L 379 | XP_005265897.1 | |
XM_005265841.4 | 1553 | Silent Mutation | CTA,CTG | L,L 379 | XP_005265898.1 | |
XM_005265842.4 | 1553 | Silent Mutation | CTA,CTG | L,L 298 | XP_005265899.1 | |
XM_005265843.2 | 1553 | Silent Mutation | CTA,CTG | L,L 298 | XP_005265900.1 | |
XM_005265844.3 | 1553 | Silent Mutation | CTA,CTG | L,L 298 | XP_005265901.1 | |
XM_005265845.4 | 1553 | Silent Mutation | CTA,CTG | L,L 298 | XP_005265902.1 | |
XM_006714831.2 | 1553 | Silent Mutation | CTA,CTG | L,L 376 | XP_006714894.1 | |
XM_006714832.2 | 1553 | Silent Mutation | CTA,CTG | L,L 332 | XP_006714895.1 | |
XM_011534466.2 | 1553 | Silent Mutation | CTA,CTG | L,L 379 | XP_011532768.1 | |
XM_011534467.2 | 1553 | Silent Mutation | CTA,CTG | L,L 298 | XP_011532769.1 |