Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCCTGCTGCCCCCTCACCTGG[T/G]GTGGGTCCTCATAACTCTCCCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4E1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4E1B - eukaryotic translation initiation factor 4E family member 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4281 - microRNA 4281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNCB - synuclein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001502.2 | 659 | Missense Mutation | CAC,CCC | H,P 123 | NP_001001502.1 | |
NM_001318034.1 | 659 | Missense Mutation | CAC,CCC | H,P 109 | NP_001304963.1 | |
NM_001318035.1 | 659 | Intron | NP_001304964.1 | |||
NM_001318036.1 | 659 | Missense Mutation | CAC,CCC | H,P 109 | NP_001304965.1 | |
NM_001318037.1 | 659 | Intron | NP_001304966.1 | |||
NM_003085.4 | 659 | Missense Mutation | CAC,CCC | H,P 123 | NP_003076.1 | |
XM_006714914.3 | 659 | Missense Mutation | CAC,CCC | H,P 123 | XP_006714977.1 | |
XM_006714915.3 | 659 | Missense Mutation | CAC,CCC | H,P 123 | XP_006714978.1 | |
XM_006714916.2 | 659 | Missense Mutation | CAC,CCC | H,P 123 | XP_006714979.1 | |
XM_011534640.2 | 659 | Missense Mutation | CAC,CCC | H,P 123 | XP_011532942.1 |