Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGTATGCGTTATTTGATGTTG[A/G]ATAAGAGCTGAACTTTGGCTAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724657 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724657 - uncharacterized LOC102724657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF354A - zinc finger protein 354A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324339.1 | 1479 | Silent Mutation | ATC,ATT | I,I 216 | NP_001311268.1 | |
NM_005649.2 | 1479 | Silent Mutation | ATC,ATT | I,I 258 | NP_005640.2 | |
XM_011534645.2 | 1479 | Silent Mutation | ATC,ATT | I,I 216 | XP_011532947.1 | |
XM_017009791.1 | 1479 | Silent Mutation | ATC,ATT | I,I 258 | XP_016865280.1 |