Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGGCACCTTGCTCCCTTCAGCG[A/G]AAGACAGGTGCCCAGGGAATGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS2 - ADAM metallopeptidase with thrombospondin type 1 motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014244.4 | 6039 | UTR 3 | NP_055059.2 | |||
NM_021599.3 | 6039 | Intron | NP_067610.1 | |||
XM_017010063.1 | 6039 | UTR 3 | XP_016865552.1 |