Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTGAAGAAGTTGACCCTATCT[G/T]CTGCAAAATAACATGAGTATATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTC-338M12.4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTC-338M12.4 - uncharacterized LOC101928649 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM52 - tripartite motif containing 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032765.2 | 1530 | UTR 3 | NP_116154.1 | |||
XM_005266000.3 | 1530 | Missense Mutation | AAG,CAG | K,Q 272 | XP_005266057.1 | |
XM_017009991.1 | 1530 | Intron | XP_016865480.1 | |||
XM_017009992.1 | 1530 | Intron | XP_016865481.1 | |||
XM_017009993.1 | 1530 | UTR 3 | XP_016865482.1 | |||
XM_017009994.1 | 1530 | UTR 3 | XP_016865483.1 |
TRIM52-AS1 - TRIM52 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |