Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCATTTCTTAAGACAGCCTGC[A/G]TGGAAAAAATGACTGCAAGGCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF145 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF145 - ring finger protein 145 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199380.1 | 1884 | Silent Mutation | CAC,CAT | H,H 587 | NP_001186309.1 | |
NM_001199381.1 | 1884 | Silent Mutation | CAC,CAT | H,H 574 | NP_001186310.1 | |
NM_001199382.1 | 1884 | Silent Mutation | CAC,CAT | H,H 571 | NP_001186311.1 | |
NM_001199383.1 | 1884 | Silent Mutation | CAC,CAT | H,H 557 | NP_001186312.1 | |
NM_144726.2 | 1884 | Silent Mutation | CAC,CAT | H,H 585 | NP_653327.1 | |
XM_017009138.1 | 1884 | Silent Mutation | CAC,CAT | H,H 557 | XP_016864627.1 |